siRNA Details
| Gene Symbol: CSNK1G2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001319.6 | NP_001310.3 | 6 | 1004 | ||
| XM_005259498.1 | XP_005259555.1 | 5 | 1106 | |||
| XM_005259499.3 | XP_005259556.1 | 5 | 913 | |||
| XM_005259500.1 | XP_005259557.1 | 6 | 1016 | |||
| XM_005259501.2 | XP_005259558.1 | 6 | 1390 | |||
| XM_017026297.1 | XP_016881786.1 | 5 | 980 | |||
| XM_017026298.1 | XP_016881787.1 | 6 | 1390 | |||
| GenBank mRNA | AB451278.1 | - | 5 | 481 | ||
| AB451410.1 | - | 5 | 481 | |||
| AF001177.1 | - | 6 | 1726 | |||
| BC018693.2 | - | 6 | 977 | 3010717 | ||
| BC018699.2 | - | 6 | 977 | 3010717 | ||
| BC020972.1 | - | 5 | 534 | 3851613 | ||
| BT009922.1 | - | 5 | 481 | |||
| U89896.1 | - | 5 | 720 | |||

CSNK1G2
Genomic Map