siRNA Details
| Gene Symbol: FABP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001444.2 | NP_001435.1 | 4 | 652 | ||
| GenBank mRNA | AK226036.1 | - | 4 | 601 | ||
| BC019385.2 | - | 4 | 585 | 3862100 | ||
| BC070303.1 | - | 1 | 599 | 6496304 | ||
| M94856.1 | - | 4 | 587 | |||

FABP5
Genomic Map