siRNA Details
| Gene Symbol: RAB3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002866.4 | NP_002857.1 | 2 | 365 | ||
| XM_011528164.1 | XP_011526466.1 | 2 | 362 | |||
| GenBank mRNA | AF254795.1 | - | 2 | 325 | ||
| AF498931.1 | - | 1 | 196 | |||
| AK289559.1 | - | 2 | 343 | |||
| AK293467.1 | - | 339 | ||||
| BC011782.2 | - | 2 | 330 | 3939176 | ||
| M28210.1 | - | 2 | 235 | |||
| X13900.1 | - | 2 | 235 | |||

RAB3A
Genomic Map