siRNA Details
| Gene Symbol: ZNF85 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256171.1 | NP_001243100.1 | 5 | 819 | ||
| NM_001256173.1 | NP_001243102.1 | 3 | 602 | |||
| NM_003429.4 | NP_003420.2 | 4 | 729 | |||
| XM_011528263.2 | XP_011526565.1 | 6 | 890 | |||
| XM_011528264.1 | XP_011526566.1 | 2 | 1050 | |||
| XM_017027234.1 | XP_016882723.1 | 2 | 529 | |||
| GenBank mRNA | AK308258.1 | - | 4 | 559 | ||
| BC047646.1 | - | 2 | 1032 | 4828290 | ||
| D70831.1 | - | 1 | 128 | |||
| U35376.1 | - | 4 | 699 | |||

ZNF85
Genomic Map