siRNA Details
| Gene Symbol: SUCLG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003848.3 | NP_003839.2 | 11 | 1651 | ||
| XM_017007420.1 | XP_016862909.1 | 11 | 2293 | |||
| GenBank mRNA | AF131748.1 | - | 5 | 924 | ||
| AL050226.1 | - | 5 | 912 | |||
| BC007716.2 | - | 11 | 1614 | 4303307 | ||
| BC019868.1 | - | 8 | 1290 | |||
| BC035149.1 | - | 10 | 1413 | |||
| BC047024.1 | - | 12 | 1621 | 5415743 | ||
| BC068602.1 | - | 11 | 1652 | 5274341 | ||

SUCLG2
Genomic Map