siRNA Details
| Gene Symbol: CNGA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005140.1 | NP_005131.1 | 7 | 1816 | ||
| GenBank mRNA | AK128186.1 | - | 7 | 1816 | ||
| BC126302.1 | - | 7 | 1685 | 8992018 | ||
| BC126304.1 | - | 7 | 1685 | 8992020 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CNGA2| Gene Symbol: CNGA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005140.1 | NP_005131.1 | 7 | 1816 | ||
| GenBank mRNA | AK128186.1 | - | 7 | 1816 | ||
| BC126302.1 | - | 7 | 1685 | 8992018 | ||
| BC126304.1 | - | 7 | 1685 | 8992020 | ||