siRNA Details
| Gene Symbol: SPRR1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199828.1 | NP_001186757.1 | 2 | 550 | ||
| NM_005987.3 | NP_005978.2 | 2 | 536 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPRR1A| Gene Symbol: SPRR1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199828.1 | NP_001186757.1 | 2 | 550 | ||
| NM_005987.3 | NP_005978.2 | 2 | 536 | |||