siRNA Details
| Gene Symbol: SPRR2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005988.2 | NP_005979.1 | 2 | 307 | ||
| GenBank mRNA | BC096108.1 | - | 1 | 280 | 40002232 | |
| BC096109.1 | - | 1 | 280 | 40002234 | ||
| BC096110.1 | - | 1 | 280 | 40002235 | ||
| BC096111.1 | - | 1 | 280 | 40002236 | ||
| BC128049.1 | - | 2 | 290 | 40115066 | ||

SPRR2A
Genomic Map