siRNA Details
| Gene Symbol: HLA-DMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006120.3 | NP_006111.2 | 3 | 674 | ||
| GenBank mRNA | AK291858.1 | - | 3 | 610 | ||
| AK298081.1 | - | 3 | 1252 | |||
| BC011447.1 | - | 3 | 647 | 4245221 | ||
| BC026279.1 | - | 3 | 603 | 4703588 | ||
| CR450324.1 | - | 3 | 588 | |||
| X62744.1 | - | 3 | 633 | |||

HLA-DMA
Genomic Map