siRNA Details
| Gene Symbol: TRIM16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006470.3 | NP_006461.3 | 4 | 816 | ||
| GenBank mRNA | AF096870.1 | - | 1 | 486 | ||
| AK002035.1 | - | 1 | 282 | |||
| AK314557.1 | - | 1 | 296 | |||
| BC001564.1 | - | 1 | 146 | |||
| BC053514.1 | - | 4 | 816 | 6263729 | ||
| BC067096.1 | - | 4 | 816 | 6088764 | ||
| CR457136.1 | - | 1 | 259 | |||

TRIM16
Genomic Map