siRNA Details
| Gene Symbol: RFPL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098535.1 | NP_001092005.1 | 2 | 1383 | ||
| NM_006604.2 | NP_006595.1 | 2 | 1109 | |||
| GenBank mRNA | AJ010232.1 | - | 2 | 1383 | ||
| AK097738.1 | - | 3 | 1861 | |||
| BC031689.1 | - | 2 | 1109 | 5167710 | ||

RFPL3
Genomic Map