siRNA Details
| Gene Symbol: ZNF17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006959.2 | NP_008890.2 | 2 | 356 | ||
| XM_017027205.1 | XP_016882694.1 | 3 | 2306 | |||
| GenBank mRNA | AK098183.1 | - | 2 | 338 | ||
| AK127948.1 | - | 3 | 395 | |||
| DA746547.1 | - | 3 | 399 | |||

ZNF17
Genomic Map