siRNA Details
| Gene Symbol: CYP4F8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007253.3 | NP_009184.1 | 7, 8 | 986 | ||
| XM_017026232.1 | XP_016881721.1 | 6, 7 | 923 | |||
| GenBank mRNA | AF133298.1 | - | 6, 7 | 929 | ||
| AK096809.1 | - | 3, 4 | 2640 | |||
| AK300530.1 | - | 6, 7 | 1077 | |||
| AK300539.1 | - | 3, 4 | 537 | |||
| AK314685.1 | - | 7, 8 | 984 | |||

CYP4F8
Genomic Map