siRNA Details
| Gene Symbol: ARHGAP21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020824.3 | NP_065875.3 | 16 | 3812 | ||
| XM_005252542.3 | XP_005252599.1 | 16 | 3880 | |||
| XM_005252544.4 | XP_005252601.1 | 15 | 5535 | |||
| XM_011519602.2 | XP_011517904.1 | 17 | 5667 | |||
| XM_011519603.2 | XP_011517905.1 | 17 | 4074 | |||
| XM_011519604.2 | XP_011517906.1 | 17 | 3890 | |||
| XM_011519605.2 | XP_011517907.1 | 16 | 5637 | |||
| XM_011519606.2 | XP_011517908.1 | 14 | 5532 | |||
| XM_011519607.2 | XP_011517909.1 | 17 | 5667 | |||
| XM_017016459.1 | XP_016871948.1 | 16 | 3972 | |||
| XM_017016460.1 | XP_016871949.1 | 15 | 3942 | |||
| XM_017016461.1 | XP_016871950.1 | 15 | 3753 | |||
| GenBank mRNA | AB037845.2 | - | 15 | 3285 | ||
| AF480466.1 | - | 16 | 3759 | |||
| AK090642.1 | - | 8 | 2027 | |||
| AK295649.1 | - | 9 | 2902 | |||
| AK307604.1 | - | 16 | 3789 | |||
| AL834495.1 | - | 8 | 1416 | |||
| BX537570.1 | - | 8 | 2814 | |||

ARHGAP21
Genomic Map