siRNA Details
| Gene Symbol: WDFY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020830.4 | NP_065881.1 | 12 | 3524 | ||
| GenBank mRNA | AB037856.1 | - | 12 | 3438 | ||
| AK022888.1 | - | 12 | 3448 | |||
| BC040525.1 | - | 12 | 3436 | 5285567 | ||
| BC065934.1 | - | 12 | 3468 | 6164567 | ||

WDFY1
Genomic Map