siRNA Details
| Gene Symbol: UBE3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014671.2 | NP_055486.2 | 16, 17 | 2408 | ||
| XM_005249564.4 | XP_005249621.1 | 15, 16 | 2342 | |||
| XM_017012818.1 | XP_016868307.1 | 17, 18 | 2680 | |||
| GenBank mRNA | AK127280.1 | - | 1, 2 | 2679 | ||
| AK295138.1 | - | 16, 17 | 2088 | |||
| D13635.1 | - | 16, 17 | 2399 | |||

UBE3C
Genomic Map