siRNA Details
| Gene Symbol: SWAP70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297714.1 | NP_001284643.1 | 11 | 2757 | ||
| NM_015055.3 | NP_055870.2 | 12 | 2931 | |||
| XM_005252830.4 | XP_005252887.1 | 10 | 2832 | |||
| GenBank mRNA | AB014540.1 | - | 12 | 2883 | ||
| AF134894.1 | - | 12 | 2901 | |||
| AK027131.1 | - | 9 | 2358 | |||

SWAP70
Genomic Map