siRNA Details
| Gene Symbol: SLC8A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015063.2 | NP_055878.1 | 5 | 2101 | ||
| XM_005259172.1 | XP_005259229.1 | 5 | 1910 | |||
| XM_017027159.1 | XP_016882648.1 | 2 | 249 | |||
| GenBank mRNA | AB029010.1 | - | 5 | 1916 | ||
| AK293457.1 | - | 5 | 842 | |||
| AK293836.1 | - | 3 | 314 | |||
| AK293860.1 | - | 4 | 1234 | |||
| AK302552.1 | - | 5 | 2101 | |||
| AL512764.1 | - | 4 | 1211 | |||

SLC8A2
Genomic Map