siRNA Details
| Gene Symbol: PYGO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015617.2 | NP_056432.1 | 3 | 430 | ||
| XM_005254300.4 | XP_005254357.1 | 3 | 551 | |||
| XM_011521446.2 | XP_011519748.1 | 3 | 825 | |||
| GenBank mRNA | AF457207.1 | - | 3 | 430 | ||
| EF625686.1 | - | 4 | 555 | |||

PYGO1
Genomic Map