siRNA Details
| Gene Symbol: GLRX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243399.1 | NP_001230328.1 | 2 | 457 | ||
| NM_001319291.1 | NP_001306220.1 | 2 | 38 | |||
| NM_016066.4 | NP_057150.2 | 2 | 615 | |||
| NM_197962.2 | NP_932066.1 | 2 | 232 | |||
| XM_017001397.1 | XP_016856886.1 | 2 | 165 | |||
| GenBank mRNA | AF132495.2 | - | 2 | 147 | ||
| AF151891.1 | - | 2 | 122 | |||
| AF290514.1 | - | 2 | 615 | |||
| AY038988.1 | - | 2 | 154 | |||
| BC009669.1 | - | 2 | 147 | |||
| BC028113.1 | - | 2 | 138 | 5395272 | ||
| BM887926.1 | - | 2 | 38 | |||
| BQ432881.1 | - | 2 | 463 | |||
| BU157087.1 | - | 187 | ||||
| CN362908.1 | - | 2 | 256 | |||

GLRX2
Genomic Map