siRNA Details
Gene Symbol: MIA-RAB4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037775.1 | 4 | 448 | |||
GenBank mRNA | AF087861.1 | - | 3 | 229 | ||
AK302147.1 | - | 5 | 864 |
Gene Symbol: RAB4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016154.4 | NP_057238.3 | 2 | 243 | ||
GenBank mRNA | AF165522.1 | - | 2 | 209 | ||
AF217985.1 | - | 1 | 1047 | |||
AF498935.1 | - | 2 | 86 | |||
BC033081.1 | - | 2 | 154 | 5456088 | ||
BC046927.1 | - | 2 | 196 | 5805921 | ||
BG706803.1 | - | 2 | 251 |
Gene Symbol: RAB4B-EGLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037791.1 | 2 | 243 | |||
GenBank mRNA | AK291385.1 | - | 2 | 191 |
