siRNA Details
Gene Symbol: RAB39A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017516.2 | NP_059986.1 | 2 | 757 | ||
GenBank mRNA | AK292969.1 | - | 2 | 729 | ||
BC028064.1 | - | 2 | 681 | 5197104 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RAB39A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017516.2 | NP_059986.1 | 2 | 757 | ||
GenBank mRNA | AK292969.1 | - | 2 | 729 | ||
BC028064.1 | - | 2 | 681 | 5197104 |