siRNA Details
| Gene Symbol: HMX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018942.2 | NP_061815.2 | 2 | 1612 | ||
| GenBank mRNA | M99587.1 | - | 2 | 1592 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HMX1| Gene Symbol: HMX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018942.2 | NP_061815.2 | 2 | 1612 | ||
| GenBank mRNA | M99587.1 | - | 2 | 1592 | ||