siRNA Details
| Gene Symbol: UGT1A8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019076.4 | NP_061949.3 | 1 | 332 | ||
| GenBank mRNA | AF030310.1 | - | 1 | 269 | ||
| AF462267.1 | - | 1 | 302 | |||
| AF462268.1 | - | 1 | 332 | |||
| AK313488.1 | - | 1 | 315 | |||
| AY435143.1 | - | 1 | 269 | |||

UGT1A8
Genomic Map