siRNA Details
| Gene Symbol: UGT1A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019077.2 | NP_061950.2 | 1 | 682 | ||
| GenBank mRNA | AY435142.1 | - | 1 | 682 | ||
| BC001521.2 | - | 1 | 252 | |||
| DQ383513.1 | - | 1 | 394 | |||
| U89507.1 | - | 1 | 682 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

UGT1A7| Gene Symbol: UGT1A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019077.2 | NP_061950.2 | 1 | 682 | ||
| GenBank mRNA | AY435142.1 | - | 1 | 682 | ||
| BC001521.2 | - | 1 | 252 | |||
| DQ383513.1 | - | 1 | 394 | |||
| U89507.1 | - | 1 | 682 | |||