siRNA Details
| Gene Symbol: OPN1MW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000513.2 | NP_000504.1 | 6 | 1276 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

OPN1MW| Gene Symbol: OPN1MW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000513.2 | NP_000504.1 | 6 | 1276 | ||
Set Membership: |
|