siRNA Details
| Gene Symbol: HPR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020995.3 | NP_066275.3 | 5 | 720 | ||
| GenBank mRNA | BM547990.1 | - | 3 | 607 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HPR| Gene Symbol: HPR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020995.3 | NP_066275.3 | 5 | 720 | ||
| GenBank mRNA | BM547990.1 | - | 3 | 607 | ||