siRNA Details
| Gene Symbol: SSX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021015.3 | NP_066295.3 | 7 | 614 | ||
| NM_175723.1 | NP_783729.1 | 6 | 491 | |||
| XM_011543949.2 | XP_011542251.1 | 6 | 526 | |||
| XM_017029758.1 | XP_016885247.1 | 6 | 526 | |||
| GenBank mRNA | BC016640.2 | - | 6 | 580 | 3923927 | |
| BE896093.1 | - | 6 | 594 | |||
| U90842.1 | - | 5 | 438 | |||

SSX5
Genomic Map