siRNA Details
| Gene Symbol: ZNF253 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021047.2 | NP_066385.2 | 4 | 510 | ||
| GenBank mRNA | AF038951.1 | - | 3 | 418 | ||
| AK074757.1 | - | 4 | 502 | |||
| AK074872.1 | - | 1 | 160 | |||
| BC014148.2 | - | 4 | 510 | |||
| BC065572.1 | - | 2 | 285 | 6171141 | ||
| BC125063.1 | - | 4 | 489 | 40117279 | ||
| BC125064.1 | - | 2 | 266 | 40117289 | ||

ZNF253
Genomic Map