siRNA Details
| Gene Symbol: CYP4F11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128932.1 | NP_001122404.1 | 13 | 1956 | ||
| NM_021187.3 | NP_067010.3 | 12 | 2378 | |||
| GenBank mRNA | AK125714.1 | - | 9 | 2083 | ||
| AL833940.1 | - | 11 | 1863 | |||

CYP4F11
Genomic Map