siRNA Details
| Gene Symbol: FXYD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022006.1 | NP_071289.1 | 3 | 193 | ||
| GenBank mRNA | AI929519.1 | - | 3 | 193 | ||
| AK057825.1 | - | 3 | 168 | |||
| BC018619.1 | - | 3 | 177 | 3626060 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FXYD7| Gene Symbol: FXYD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022006.1 | NP_071289.1 | 3 | 193 | ||
| GenBank mRNA | AI929519.1 | - | 3 | 193 | ||
| AK057825.1 | - | 3 | 168 | |||
| BC018619.1 | - | 3 | 177 | 3626060 | ||