siRNA Details
| Gene Symbol: ZXDC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025112.4 | NP_079388.3 | 10 | 2764 | ||
| NR_104249.1 | 9 | 2944 | ||||
| XM_006713741.2 | XP_006713804.1 | 8 | 2509 | |||
| XM_011513119.2 | XP_011511421.1 | 9 | 2612 | |||
| GenBank mRNA | AK023923.1 | - | 7 | 1466 | ||
| AK098501.1 | - | 2 | 3998 | |||
| AL390142.1 | - | 2 | 1980 | |||
| AY936556.1 | - | 10 | 2724 | |||
| BC002940.2 | - | 9 | 1702 | |||
| BC012729.2 | - | 9 | 1702 | |||
| BC063713.1 | - | 9 | 2012 | |||
| DQ079590.1 | - | 10 | 2764 | |||

ZXDC
Genomic Map