siRNA Details
| Gene Symbol: ATAD3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK127857.1 | - | 3233 | |||
| BC002542.2 | - | 2388 | 3138578 | |||
| BC018701.1 | - | 2388 | 3139886 | |||
| Gene Symbol: ATAD3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK025865.1 | - | 1500 | |||
| BC007803.2 | - | 2461 | 4136170 | |||
| BC014101.2 | - | 2490 | 3503438 | |||
| BC033109.1 | - | 2464 | 4877449 | |||

ATAD3B
Genomic Map