siRNA Details
| Gene Symbol: ADGRA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005252695.1 | XP_005252752.1 | 3 | 437 | ||
| GenBank mRNA | AY181241.1 | - | 3 | 437 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ADGRA1| Gene Symbol: ADGRA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005252695.1 | XP_005252752.1 | 3 | 437 | ||
| GenBank mRNA | AY181241.1 | - | 3 | 437 | ||