siRNA Details
| Gene Symbol: NT5C1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032526.2 | NP_115915.1 | 4 | 511 | ||
| GenBank mRNA | AF331801.1 | - | 4 | 511 | ||
| AY028778.1 | - | 4 | 502 | |||
| BC103879.1 | - | 4 | 511 | 40004489 | ||
| BC103880.1 | - | 4 | 511 | 40004491 | ||

NT5C1A
Genomic Map