siRNA Details
| Gene Symbol: ZNF682 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077349.1 | NP_001070817.1 | 3 | 249 | ||
| NM_033196.2 | NP_149973.1 | 3 | 292 | |||
| XM_017027455.1 | XP_016882944.1 | 2 | 396 | |||
| XM_017027456.1 | XP_016882945.1 | 3 | 287 | |||
| GenBank mRNA | AK096571.1 | - | 3 | 290 | ||
| AK098164.1 | - | 5 | 495 | |||
| BC136304.1 | - | 2 | 209 | 9020291 | ||
| DA612976.1 | - | 3 | 294 | |||
| DA788529.1 | - | 3 | 288 | |||
| DB174863.1 | - | 3 | 270 | |||
| DR002650.1 | - | 3 | 262 | |||

ZNF682
Genomic Map