siRNA Details
| Gene Symbol: ZNF257 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001316996.1 | NP_001303925.1 | 5 | 2426 | ||
| NM_001316997.1 | NP_001303926.1 | 5 | 2370 | |||
| NM_001316998.1 | NP_001303927.1 | 2 | 2082 | |||
| NM_033468.3 | NP_258429.2 | 4 | 2305 | |||
| GenBank mRNA | AF070651.1 | - | 5 | 2120 | ||
| AK056694.1 | - | 4 | 2190 | |||
| BC036446.1 | - | 5 | 2420 | 4831021 | ||

ZNF257
Genomic Map