siRNA Details
| Gene Symbol: MYL12B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144944.1 | NP_001138416.1 | 4 | 755 | ||
| NM_001144945.1 | NP_001138417.1 | 4 | 677 | |||
| NM_033546.3 | NP_291024.1 | 4 | 736 | |||
| GenBank mRNA | AK291726.1 | - | 4 | 716 | ||
| AY320408.1 | - | 4 | 740 | |||
| BC004994.1 | - | 4 | 655 | 2906108 | ||
| CN302451.1 | - | 4 | 658 | |||
| D50372.1 | - | 4 | 688 | |||
| D82058.1 | - | 4 | 669 | |||
| U26162.1 | - | 4 | 710 | |||

MYL12B
Genomic Map