siRNA Details
| Gene Symbol: ZNF721 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133474.3 | NP_597731.2 | 3 | 648 | ||
| GenBank mRNA | AK127067.1 | - | 1 | 204 | ||
| AK128499.1 | - | 1 | 477 | |||
| AL834155.1 | - | 452 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF721| Gene Symbol: ZNF721 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133474.3 | NP_597731.2 | 3 | 648 | ||
| GenBank mRNA | AK127067.1 | - | 1 | 204 | ||
| AK128499.1 | - | 1 | 477 | |||
| AL834155.1 | - | 452 | ||||