siRNA Details
| Gene Symbol: ZNF513 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201459.1 | NP_001188388.1 | 3 | 1760 | ||
| NM_144631.5 | NP_653232.3 | 4 | 1837 | |||
| XM_005264142.2 | XP_005264199.1 | 4 | 1670 | |||
| XM_005264143.3 | XP_005264200.1 | 3 | 1224 | |||
| GenBank mRNA | AB075875.1 | - | 3 | 1760 | ||
| AK056765.1 | - | 2 | 1142 | |||
| AK096933.1 | - | 3 | 1699 | |||
| AK290690.1 | - | 4 | 1811 | |||
| AL833946.1 | - | 1 | 998 | |||
| BC040650.1 | - | 2 | 1131 | |||
| BC052282.1 | - | 4 | 1819 | 5467936 | ||

ZNF513
Genomic Map