siRNA Details
| Gene Symbol: DHX57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198963.1 | NP_945314.1 | 5 | 767 | ||
| XM_011533154.2 | XP_011531456.1 | 5 | 771 | |||
| XM_011533155.2 | XP_011531457.1 | 5 | 769 | |||
| XM_011533156.2 | XP_011531458.1 | 5 | 771 | |||
| XM_011533157.1 | XP_011531459.1 | 5 | 771 | |||
| GenBank mRNA | AK057423.1 | - | 4 | 567 | ||
| AK296739.1 | - | 5 | 806 | |||
| AK307514.1 | - | 5 | 796 | |||
| BC039270.1 | - | 4 | 574 | |||
| BC060778.1 | - | 5 | 767 | |||
| BC131534.1 | - | 5 | 739 | 40082400 | ||

DHX57
Genomic Map