siRNA Details
Gene Symbol: DNAJB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145174.1 | NP_660157.1 | 1 | 1674 | ||
GenBank mRNA | AF085232.1 | - | 1 | 1674 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DNAJB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145174.1 | NP_660157.1 | 1 | 1674 | ||
GenBank mRNA | AF085232.1 | - | 1 | 1674 |