siRNA Details
| Gene Symbol: GJD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152219.3 | NP_689343.3 | 1 | 3960 | ||
| GenBank mRNA | AK125254.1 | - | 2 | 2015 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GJD3| Gene Symbol: GJD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152219.3 | NP_689343.3 | 1 | 3960 | ||
| GenBank mRNA | AK125254.1 | - | 2 | 2015 | ||