siRNA Details
| Gene Symbol: MCOLN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153259.3 | NP_694991.2 | 9 | 1316 | ||
| XM_005270719.3 | XP_005270776.1 | 9 | 1093 | |||
| XM_006710552.2 | XP_006710615.1 | 10 | 1406 | |||
| XM_011541187.2 | XP_011539489.1 | 9 | 1150 | |||
| XM_011541188.2 | XP_011539490.1 | 9 | 1433 | |||
| GenBank mRNA | AK129786.1 | - | 3 | 319 | ||
| AK309858.1 | - | 9 | 1307 | |||
| AY083533.1 | - | 8 | 982 | |||
| BC104891.1 | - | 9 | 1134 | 8143894 | ||
| BC104893.1 | - | 9 | 1134 | 8143896 | ||

MCOLN2
Genomic Map