siRNA Details
| Gene Symbol: TMEM171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161342.2 | NP_001154814.1 | 4 | 1017 | ||
| NM_173490.7 | NP_775761.4 | 4 | 1020 | |||
| XM_011543156.1 | XP_011541458.1 | 4 | 943 | |||
| GenBank mRNA | BC018083.1 | - | 4 | 1006 | 4797244 | |
| BC035310.1 | - | 3 | 858 | 4746772 | ||

TMEM171
Genomic Map