siRNA Details
| Gene Symbol: PIP5KL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135219.1 | NP_001128691.1 | 7, 8 | 703 | ||
| NM_173492.1 | NP_775763.1 | 2, 3 | 259 | |||
| GenBank mRNA | BC042184.1 | - | 2, 3 | 259 | 5163193 | |
| BE881330.1 | - | 4, 5 | 286 | |||

PIP5KL1
Genomic Map