siRNA Details
| Gene Symbol: TMEM86B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173804.4 | NP_776165.3 | 3 | 1364 | ||
| XM_017026554.1 | XP_016882043.1 | 2 | 1137 | |||
| GenBank mRNA | AK126545.1 | - | 1 | 1764 | ||
| BC023000.1 | - | 3 | 1364 | 5000011 | ||
| BC033193.1 | - | 2 | 1126 | |||

TMEM86B
Genomic Map