siRNA Details
| Gene Symbol: ZNF575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174945.2 | NP_777605.1 | 4 | 1391 | ||
| XM_005258783.3 | XP_005258840.1 | 4 | 1340 | |||
| XM_011526793.2 | XP_011525095.1 | 4 | 1133 | |||
| GenBank mRNA | AK057129.1 | - | 5 | 1967 | ||
| BC043611.1 | - | 4 | 1224 | 6018699 | ||

ZNF575
Genomic Map