siRNA Details
| Gene Symbol: KRT77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175078.2 | NP_778253.2 | 9 | 2423 | ||
| XM_011538288.2 | XP_011536590.1 | 9 | 2185 | |||
| GenBank mRNA | AJ564104.1 | - | 9 | 2423 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRT77| Gene Symbol: KRT77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175078.2 | NP_778253.2 | 9 | 2423 | ||
| XM_011538288.2 | XP_011536590.1 | 9 | 2185 | |||
| GenBank mRNA | AJ564104.1 | - | 9 | 2423 | ||