siRNA Details
| Gene Symbol: ANGPTL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178127.4 | NP_835228.2 | 4 | 922 | ||
| XM_011542735.2 | XP_011541037.1 | 4 | 916 | |||
| GenBank mRNA | AK291523.1 | - | 3 | 386 | ||
| AY169281.1 | - | 3 | 380 | |||
| AY358827.1 | - | 3 | 370 | |||
| BC049170.1 | - | 4 | 925 | 5285447 | ||
| BC053957.1 | - | 4 | 925 | |||

ANGPTL5
Genomic Map